Learn More
Waters Corp HPLC Guard Cartridge, Atlantis dC18 Prep, 100 Å, 5 μm, 10 x 10 mm
Supplier:  Waters Corp 186002300
Encompass_Preferred
This HPLC Guard Cartridge, Atlantis dC18 Prep, is designed for high-performance liquid chromatography. It features 100 Å, 5 μm media and dimensions of 10 x 10 mm. This cartridge helps extend the life of your analytical column by protecting it from particulate matter and irreversibly adsorbed sample components.
  • High-value analytical technologies
  • Industry-leading scientific expertise
  • Innovations in chromatography, mass spectrometry, and thermal analysis
  • High-quality instruments
  • HPLC separation columns
  • GC separation columns
  • Used for discovering new pharmaceuticals
  • Used for assuring the safety of the world's food and water supplies
  • Used for ensuring the integrity of a chemical entity in production
  • Full range of highest quality liquid chromatography products and services
  • Competitive prices
Catalog No. 50-781-061