Learn More
Waters Corp Atlantis dC18 Prep Column, 100Å, 10 µm, 10 mm X 10 mm, 1/pk
Catalog No. 50781118 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Atlantis dC18 columns are universal, silica-based, reversed-phase C18 columns that are used for polar compound retention. The Atlantis dC18 columns feature di-functionally bonded C18 ligands that have been optimized for use with highly aqueous mobile phases, including 100% water.
Catalog No. 50-781-118 Supplier Waters Corp Supplier No. 186002452
Add to Cart
Add to Cart
Atlantis dC18 columns are universal, silica-based, reversed-phase C18 columns that are used for polar compound retention. The Atlantis dC18 columns feature di-functionally bonded C18 ligands that have been optimized for use with highly aqueous mobile phases, including 100% water.