Learn More
Waters Corp Atlantis dC18 OBD Prep Column, 100Å, 5 µm, 30 mm X 150 mm, 1/pk
Catalog No. 50781369 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Atlantis dC18 columns are universal, silica-based, reversed-phase C18 columns that are used for polar compound retention. The Atlantis dC18 columns feature di-functionally bonded C18 ligands that have been optimized for use with highly aqueous mobile phases, including 100% water.
Catalog No. 50-781-369 Supplier Waters Corp Supplier No. 186002801
Add to Cart
Add to Cart
Request a Quote
Atlantis dC18 columns are universal, silica-based, reversed-phase C18 columns that are used for polar compound retention. The Atlantis dC18 columns feature di-functionally bonded C18 ligands that have been optimized for use with highly aqueous mobile phases, including 100% water.