Learn More
Waters Corp WATERS CORPORATION Preparative Column, Atlantis dC18, 100Å, 5 μm, 19 mm × 150 mm, OBD Prep
Supplier:  Waters Corp 186002800
Encompass_Preferred
Atlantis dC18 columns are universal, silica-based, reversed-phase C18 columns used for polar compound retention. They feature di-functionally bonded C18 ligands optimized for use with highly aqueous mobile phases, including 100% water.
  • Universal, silica-based, reversed-phase C18 column.
  • Used for polar compound retention.
  • Features di-functionally bonded C18 ligands optimized for use with highly aqueous mobile phases, including 100% water.
Catalog No. 50-781-368