Learn More
Waters Corp HILIC OBD Prep Column, Silica, 100Å, 5 μm, 19 mm × 50 mm
Supplier:  Waters Corp 186003957
Encompass_Preferred
HILIC Silica columns are designed for the retention of polar analytes in high organic mobile phases, typically greater than 80% acetonitrile. These columns provide an orthogonal analyte selectivity compared to traditional reversed-phase chromatography, making them suitable for compounds that are difficult to retain with conventional methods.
  • Designed for use with high organic mobile phases, typically >80% acetonitrile.
  • Provides retention for polar analytes.
  • Offers orthogonal analyte selectivity compared to traditional reversed-phase approaches.
  • Suitable for compounds not retained by reversed-phase chromatography.
Catalog No. 50-782-087