Learn More
Waters Corp WATERS CORPORATION Atlantis HILIC OBD Prep Column, 30 mm x 100 mm, 5 μm, 100 Å
Catalog No. 50782091 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Atlantis HILIC columns provide robust retention for polar, basic, water-soluble analytes. Optimized for HILIC separations, they feature traditional stainless steel OBD technology, excellent pH stability, and low MS bleed.
  • Excellent retention for polar, basic, water-soluble analytes.
  • Optimized for HILIC separations with high organic mobile phases.
  • Traditional stainless steel OBD column technology.
  • Excellent low and high pH stability.
  • Minimizes MS bleed.
  • Uniform, optimum packed bed density for Prep LC.
  • Direct scalability from analytical to preparative chromatography.
  • Provides excellent column lifetime and consistent performance.
  • Delivers good resolution, peak shape, and mass loading.
Catalog No. 50-782-091 Supplier Waters Corp Supplier No. 186003961
Add to Cart
Add to Cart
Request a Quote
Atlantis HILIC columns provide robust retention for polar, basic, water-soluble analytes. Optimized for HILIC separations, they feature traditional stainless steel OBD technology, excellent pH stability, and low MS bleed.
  • Excellent retention for polar, basic, water-soluble analytes.
  • Optimized for HILIC separations with high organic mobile phases.
  • Traditional stainless steel OBD column technology.
  • Excellent low and high pH stability.
  • Minimizes MS bleed.
  • Uniform, optimum packed bed density for Prep LC.
  • Direct scalability from analytical to preparative chromatography.
  • Provides excellent column lifetime and consistent performance.
  • Delivers good resolution, peak shape, and mass loading.