Learn More
Waters Corp Atlantis T3 OBD Prep Column, 100Å, 10 μm, 30 mm × 75 mm
Catalog No. 50466041 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Atlantis T3 columns are universal, silica-based, reversed-phase C18 columns that provide balanced retention of polar and hydrophobic molecules and should be your first choice when developing separations of polar and non-polar compounds.
  • Universal, silica-based, reversed-phase C18 columns.
  • Provide balanced retention of polar and hydrophobic molecules.
  • Recommended as a first choice for developing separations of polar and non-polar compounds.
Catalog No. 50-466-041 Supplier Waters Corp Supplier No. 186004712
Add to Cart
Add to Cart
Request a Quote
Atlantis T3 columns are universal, silica-based, reversed-phase C18 columns that provide balanced retention of polar and hydrophobic molecules and should be your first choice when developing separations of polar and non-polar compounds.
  • Universal, silica-based, reversed-phase C18 columns.
  • Provide balanced retention of polar and hydrophobic molecules.
  • Recommended as a first choice for developing separations of polar and non-polar compounds.