Learn More
Waters Corp Waters Prep Column, Atlantis T3, 100Å, 10 μm, 50 × 100 mm
Catalog No. 50782168 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This silica column is designed for preparative HPLC applications. Featuring Optimum Bed Density (OBD) technology, it provides uniform density profiles, exceptional resistance to mechanical failure, and consistent column performance. This design maximizes purification productivity by facilitating method scaling and extending column lifetimes.
  • Silica column for polar retention.
  • Optimum Bed Density (OBD) packing for predictable, uniform density profiles.
  • Exceptional resistance to mechanical chromatographic bed failure.
  • Consistent column-to-column performance.
  • Extended column lifetimes reduce costs.
  • Maximizes productivity in purification.
  • Facilitates scaling up of screening methods to preparative HPLC.
  • Increases laboratory throughput when scaling methods from UPLC to OBD Preparative HPLC Columns.
Catalog No. 50-782-168 Supplier Waters Corp Supplier No. 186004084
Add to Cart
Add to Cart
Request a Quote
This silica column is designed for preparative HPLC applications. Featuring Optimum Bed Density (OBD) technology, it provides uniform density profiles, exceptional resistance to mechanical failure, and consistent column performance. This design maximizes purification productivity by facilitating method scaling and extending column lifetimes.
  • Silica column for polar retention.
  • Optimum Bed Density (OBD) packing for predictable, uniform density profiles.
  • Exceptional resistance to mechanical chromatographic bed failure.
  • Consistent column-to-column performance.
  • Extended column lifetimes reduce costs.
  • Maximizes productivity in purification.
  • Facilitates scaling up of screening methods to preparative HPLC.
  • Increases laboratory throughput when scaling methods from UPLC to OBD Preparative HPLC Columns.