Learn More
Waters Corp Waters Corporation OBD Prep Column, C18, 5 μm, 50 mm X 50 mm, 100 Å
Catalog No. 50782164 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Atlantis T3 columns are universal, silica-based, reversed-phase C18 columns that offer balanced retention for both polar and hydrophobic molecules. They are recommended as a primary choice for developing separations of polar and non-polar compounds.
  • Polar compound retention
  • 100% Aqueous compatibility
  • Long column life at low pH
  • High-mass loading
  • Low-pH stability
  • Excellent column life and stability
  • Superior peak shapes
Catalog No. 50-782-164 Supplier Waters Corp Supplier No. 186004080
Add to Cart
Add to Cart
Request a Quote
Atlantis T3 columns are universal, silica-based, reversed-phase C18 columns that offer balanced retention for both polar and hydrophobic molecules. They are recommended as a primary choice for developing separations of polar and non-polar compounds.
  • Polar compound retention
  • 100% Aqueous compatibility
  • Long column life at low pH
  • High-mass loading
  • Low-pH stability
  • Excellent column life and stability
  • Superior peak shapes