Learn More
Waters Corp Atlantis Silica HILIC Column, 100Å, 3 μm, 4.6 mm X 150 mm
Catalog No. 502392510 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Atlantis Silica HILIC Column with eConnect Column Tag utilizes smart-column technology to enhance compliance and data management in laboratories. Its electronic tag stores product details, facilitates data integration with Empower CDS, and offers NFC access to product information.
  • Simplifies setup and reduces manual data entry with pre-loaded column-specific information.
  • Enhances compliance and system setup with a permanent, tamper-proof tag, aiding auto-detectability.
  • Provides NFC-capability for easy access to column-specific information and knowledge.
  • Offers identical performance and quality standards to untagged counterparts.
Catalog No. 50-239-2510 Supplier Waters Corp Supplier No. 186002031RF
The Atlantis Silica HILIC Column with eConnect Column Tag utilizes smart-column technology to enhance compliance and data management in laboratories. Its electronic tag stores product details, facilitates data integration with Empower CDS, and offers NFC access to product information.
  • Simplifies setup and reduces manual data entry with pre-loaded column-specific information.
  • Enhances compliance and system setup with a permanent, tamper-proof tag, aiding auto-detectability.
  • Provides NFC-capability for easy access to column-specific information and knowledge.
  • Offers identical performance and quality standards to untagged counterparts.