Learn More
Waters Corp VanGuard Cartridge, Silica HILIC, 100Å, 5μm, 2.1 mm x 5 mm
Catalog No. 502391635 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
VanGuard Cartridges are designed to extend analytical column lifetime and performance by removing particulate contamination from the mobile phase stream. These cartridges are optimized to protect all 2.1 mm and 3.0 mm ID HILIC analytical columns that contain 5 μm sorbent particles.
  • Extends analytical column lifetime.
  • Improves column performance.
  • Removes particulate contamination from the mobile phase stream.
  • Optimized for 2.1 mm and 3.0 mm ID HILIC analytical columns.
  • Compatible with 5 μm sorbent particles.
Catalog No. 50-239-1635 Supplier Waters Corp Supplier No. 186007670
VanGuard Cartridges are designed to extend analytical column lifetime and performance by removing particulate contamination from the mobile phase stream. These cartridges are optimized to protect all 2.1 mm and 3.0 mm ID HILIC analytical columns that contain 5 μm sorbent particles.
  • Extends analytical column lifetime.
  • Improves column performance.
  • Removes particulate contamination from the mobile phase stream.
  • Optimized for 2.1 mm and 3.0 mm ID HILIC analytical columns.
  • Compatible with 5 μm sorbent particles.