Learn More
Waters Corp VanGuard Cartridge, HILIC, 100Å, 3 μm, 2.1 mm × 5 mm
Catalog No. 502391628 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This VanGuard Cartridge extends analytical column lifetime and performance by removing particulate contamination from the mobile phase stream. It is optimized to protect 2.1 mm and 3.0 mm I.D. HILIC analytical columns with 3 μm sorbent, suitable for reversed-phase liquid chromatography.
  • Extends analytical column lifetime and performance by removing particulate and chemical contamination
  • Protects 2.1 mm and 3.0 mm I.D. HILIC analytical columns with 3 μm sorbent
  • Manufactured under cGMP and ISO 9001 guidelines
  • Tested and certified for peak performance, selectivity, sensitivity, and speed
  • Operates across a 1 to 5 pH range
  • Sustains maximum pressure up to 18000 psi (1240 Bar)
  • Directly links to the inlet side of a HILIC column
Catalog No. 50-239-1628 Supplier Waters Corp Supplier No. 186007666
This VanGuard Cartridge extends analytical column lifetime and performance by removing particulate contamination from the mobile phase stream. It is optimized to protect 2.1 mm and 3.0 mm I.D. HILIC analytical columns with 3 μm sorbent, suitable for reversed-phase liquid chromatography.
  • Extends analytical column lifetime and performance by removing particulate and chemical contamination
  • Protects 2.1 mm and 3.0 mm I.D. HILIC analytical columns with 3 μm sorbent
  • Manufactured under cGMP and ISO 9001 guidelines
  • Tested and certified for peak performance, selectivity, sensitivity, and speed
  • Operates across a 1 to 5 pH range
  • Sustains maximum pressure up to 18000 psi (1240 Bar)
  • Directly links to the inlet side of a HILIC column