Learn More
Waters Corp Atlantis VanGuard Cartridge, HILIC, Silica, 100Å, 3 μm
Catalog No. 502390487 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Atlantis HILIC VanGuard Cartridges extend the lifetime and performance of analytical columns by removing particulate contamination from the mobile phase. They are optimized to protect analytical columns containing 3 μm sorbent particles and are used with high organic mobile phases (>80% acetonitrile) to retain polar analytes, offering orthogonal selectivity compared to reversed-phase methods.
  • Extends analytical column lifetime and performance.
  • Removes particulate contamination from the mobile phase stream.
  • Optimized for 3.9 mm and 4.6 mm I.D. analytical columns with 3 μm sorbent particles.
  • Retains polar analytes using high organic mobile phases (>80% acetonitrile).
  • Provides orthogonal analyte selectivity.
  • Produced in compliance with cGMP and ISO 9001 standards.
  • Ensures optimal efficiency, selectivity, sensitivity, and speed.
Catalog No. 50-239-0487 Supplier Waters Corp Supplier No. 186007668
Atlantis HILIC VanGuard Cartridges extend the lifetime and performance of analytical columns by removing particulate contamination from the mobile phase. They are optimized to protect analytical columns containing 3 μm sorbent particles and are used with high organic mobile phases (>80% acetonitrile) to retain polar analytes, offering orthogonal selectivity compared to reversed-phase methods.
  • Extends analytical column lifetime and performance.
  • Removes particulate contamination from the mobile phase stream.
  • Optimized for 3.9 mm and 4.6 mm I.D. analytical columns with 3 μm sorbent particles.
  • Retains polar analytes using high organic mobile phases (>80% acetonitrile).
  • Provides orthogonal analyte selectivity.
  • Produced in compliance with cGMP and ISO 9001 standards.
  • Ensures optimal efficiency, selectivity, sensitivity, and speed.