Learn More
Waters Corp Prep Guard Cartridge, Silica T3, 100Å, 10 μm, 30 mm X 10 mm
Catalog No. 502391368 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This guard cartridge protects HPLC and SFC 30 mm ID Prep OBD™ columns. It extends column lifetime and reduces purification costs by removing contaminants from the mobile phase stream, ensuring consistent analytical performance.
  • Protects HPLC and SFC 30 mm ID Prep OBD™ columns.
  • Extends column lifetime and reduces purification costs.
  • Compatible with T3 Reversed-Phase Liquid Chromatography Columns.
  • Uses identical packing materials for consistent performance.
  • Removes chemical and particle contaminants.
  • Maintains separation efficiency in UPLC, UHPLC, and HPLC.
  • Withstands pressures up to 6000 psi (415 Bar).
  • Suitable for pH levels from 2 to 8.
  • Manufactured under cGMP and ISO 9001 guidelines.
  • Each unit tested and certified for optimal performance.
Catalog No. 50-239-1368 Supplier Waters Corp Supplier No. 186006878
This guard cartridge protects HPLC and SFC 30 mm ID Prep OBD™ columns. It extends column lifetime and reduces purification costs by removing contaminants from the mobile phase stream, ensuring consistent analytical performance.
  • Protects HPLC and SFC 30 mm ID Prep OBD™ columns.
  • Extends column lifetime and reduces purification costs.
  • Compatible with T3 Reversed-Phase Liquid Chromatography Columns.
  • Uses identical packing materials for consistent performance.
  • Removes chemical and particle contaminants.
  • Maintains separation efficiency in UPLC, UHPLC, and HPLC.
  • Withstands pressures up to 6000 psi (415 Bar).
  • Suitable for pH levels from 2 to 8.
  • Manufactured under cGMP and ISO 9001 guidelines.
  • Each unit tested and certified for optimal performance.