Learn More
Waters Corp HPLC Column, 100Å, 3 μm, 2.1 mm x 150 mm, C18
Catalog No. 502392423 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Atlantis T3 columns are universal, silica-based, reversed-phase C18 columns that provide balanced retention of both polar and hydrophobic molecules, making them suitable for developing separations of polar and non-polar compounds.
  • Provides balanced retention for polar and hydrophobic molecules.
  • Utilizes advanced T3 bonding and proprietary T3 end-capping for superior polar compound retention and aqueous compatibility.
  • Offers enhanced column performance, lifetime, peak shape, and stability.
  • Formulated for longer lifetimes at low pH and improved peak shape for amine-containing bases at pH 7.
  • Serves as a universal silica-based, reversed-phase C18 column for a wide range of applications.
Catalog No. 50-239-2423 Supplier Waters Corp Supplier No. 186003719RF
Atlantis T3 columns are universal, silica-based, reversed-phase C18 columns that provide balanced retention of both polar and hydrophobic molecules, making them suitable for developing separations of polar and non-polar compounds.
  • Provides balanced retention for polar and hydrophobic molecules.
  • Utilizes advanced T3 bonding and proprietary T3 end-capping for superior polar compound retention and aqueous compatibility.
  • Offers enhanced column performance, lifetime, peak shape, and stability.
  • Formulated for longer lifetimes at low pH and improved peak shape for amine-containing bases at pH 7.
  • Serves as a universal silica-based, reversed-phase C18 column for a wide range of applications.