Learn More
Waters Corp Atlantis T3 Column, C18, 3 μm, 2.1 mm x 50 mm, 100 Å
Catalog No. 502390882 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Atlantis T3 columns are universal, silica-based, reversed-phase C18 columns designed for balanced retention of polar and hydrophobic molecules. They are recommended for developing separations of polar and non-polar compounds.
  • Universal, silica-based, reversed-phase C18 columns.
  • Provides balanced retention of polar and hydrophobic molecules.
  • Equipped with an electronic eConnect column tag containing unique column-specific information.
  • NFC-enabled device-tagged columns automatically identify, track, and digitally trace column use history.
  • Can be used on any HPLC.
  • Advantages of eConnect columns are unlocked with compatible systems and software.
Catalog No. 50-239-0882 Supplier Waters Corp Supplier No. 186003717RF
The Atlantis T3 columns are universal, silica-based, reversed-phase C18 columns designed for balanced retention of polar and hydrophobic molecules. They are recommended for developing separations of polar and non-polar compounds.
  • Universal, silica-based, reversed-phase C18 columns.
  • Provides balanced retention of polar and hydrophobic molecules.
  • Equipped with an electronic eConnect column tag containing unique column-specific information.
  • NFC-enabled device-tagged columns automatically identify, track, and digitally trace column use history.
  • Can be used on any HPLC.
  • Advantages of eConnect columns are unlocked with compatible systems and software.