Learn More
Waters Corp Column, C18, 5 μm, 4.6 mm x 250 mm
Supplier: Waters Corp 186003748RF
This column is equipped with an electronic eConnect column tag that contains unique column-specific information. These NFC-enabled device-tagged columns are designed to automatically identify, track, and digitally trace column use history. While eConnect columns can be used on any HPLC, their advantages are fully realized when used with the Waters Alliance iS System running Empower software.
- Equipped with an electronic eConnect column tag with unique column-specific information.
- NFC-enabled device-tagged columns automatically identify, track, and digitally trace column use history.
- The advantages of eConnect columns are fully realized when used with the Alliance iS System running Empower software.