Learn More
Waters Corp Atlantis T3 Column, C18, 5 μm, 4.6 mm × 150 mm
Supplier:  Waters Corp 186003747RF
Encompass_Preferred
The Atlantis T3 Column is a C18 reversed-phase column built on a spherical silica particle substrate. It features a 100 Å pore size, 5 μm particle size, and dimensions of 4.6 mm × 150 mm. This column is designed with T3 bonding technology and can withstand a maximum pressure of 6000 psi (415 Bar). It includes an eConnect Column Tag for automatic identification, tracking, and digital tracing of column usage.
  • Equipped with an electronic eConnect column tag for unique, traceable column-specific information.
  • NFC-enabled design allows automatic identification, tracking, and digital tracing of column use history.
  • Compatible with any HPLC, with enhanced features on compatible iS systems running Empower software.
  • Features C18 chemistry, spherical silica particles, and T3 bonding technology.
  • Withstands maximum pressure of 6000 psi (415 Bar) and offers a pH range of 2 to 8.
Catalog No. 50-239-0555