Learn More
Waters Corp Waters Corporation Column, C18, 100Å, 3 μm, 2.1 mm X 50 mm
Catalog No. 50781980 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These universal, silica-based, reversed-phase C18 columns offer balanced retention for polar and hydrophobic molecules, enabling superior separation of highly polar compounds across a wide pH range. Advanced T3 bonding and proprietary T3 end-capping enhance performance, lifetime, peak shape, and stability, making them an excellent alternative to traditional C18 reversed-phase LC columns.
  • Universal silica-based, reversed-phase C18 column offering balanced retention for polar and hydrophobic molecules and superior performance across a wide pH range with improved peak shape for amine-containing bases at pH 7
  • Longer lifetimes at low pH
  • Advanced T3 bonding and proprietary T3 end-capping processes for enhanced column performance, lifetime, peak shape, and stability
Catalog No. 50-781-980 Supplier Waters Corp Supplier No. 186003717
These universal, silica-based, reversed-phase C18 columns offer balanced retention for polar and hydrophobic molecules, enabling superior separation of highly polar compounds across a wide pH range. Advanced T3 bonding and proprietary T3 end-capping enhance performance, lifetime, peak shape, and stability, making them an excellent alternative to traditional C18 reversed-phase LC columns.
  • Universal silica-based, reversed-phase C18 column offering balanced retention for polar and hydrophobic molecules and superior performance across a wide pH range with improved peak shape for amine-containing bases at pH 7
  • Longer lifetimes at low pH
  • Advanced T3 bonding and proprietary T3 end-capping processes for enhanced column performance, lifetime, peak shape, and stability