Learn More
Waters Corp Waters Corporation Atlantis T3 Column, 100Å, 3 μm, 3 mm x 50 mm
Catalog No. 50781984 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Atlantis T3 columns are universal, silica-based, reversed-phase C18 columns providing balanced retention of polar and hydrophobic molecules. They are a primary choice for developing separations of polar and non-polar compounds. Columns offer extended longevity at low pH and improved peak shape for amine-containing bases at pH 7, ensuring enhanced performance across a broad pH range. They retain and separate highly polar chemicals, offering strong retention and selectivity for neutral and hydrophobic chemicals in addition to polar analytes.
  • Universal, silica-based, reversed-phase C18 column.
  • Balanced retention of polar & hydrophobic molecules.
  • Extended longevity; improved peak shape at low pH.
  • Effective over wide pH range (2-8 pH).
  • T3 bonding for polar retention, aqueous compatibility.
  • Patented T3 end-capping for enhanced efficiency.
  • Max pressure: 6000 psi (415 Bar).
Catalog No. 50-781-984 Supplier Waters Corp Supplier No. 186003721
Add to Cart
Add to Cart
Atlantis T3 columns are universal, silica-based, reversed-phase C18 columns providing balanced retention of polar and hydrophobic molecules. They are a primary choice for developing separations of polar and non-polar compounds. Columns offer extended longevity at low pH and improved peak shape for amine-containing bases at pH 7, ensuring enhanced performance across a broad pH range. They retain and separate highly polar chemicals, offering strong retention and selectivity for neutral and hydrophobic chemicals in addition to polar analytes.
  • Universal, silica-based, reversed-phase C18 column.
  • Balanced retention of polar & hydrophobic molecules.
  • Extended longevity; improved peak shape at low pH.
  • Effective over wide pH range (2-8 pH).
  • T3 bonding for polar retention, aqueous compatibility.
  • Patented T3 end-capping for enhanced efficiency.
  • Max pressure: 6000 psi (415 Bar).