Learn More
Waters Corp Waters Atlantis T3 Column, 100Å, 3 μm, 4.6 mm, 150 mm
Supplier:  Waters Corp 186003729
Encompass_Preferred
Atlantis T3 columns are universal, silica-based, reversed-phase C18 columns designed for balanced retention of both polar and hydrophobic molecules. They are ideal for developing separations of polar and non-polar compounds, offering superior performance, versatility, and chromatographic stability for polar compounds.
  • Universal, silica-based, reversed-phase C18 columns.
  • Provides balanced retention of polar and hydrophobic molecules.
  • Advanced T3 bonding process enhances polar compound retention and ensures exceptional column life.
  • Proprietary T3 end-capping results in superior peak shape and stability.
  • Enables lower limits of detection and quantitation for LC/MS methods.
  • 3 μm particle size enables high-resolution separations with reduced run times.
  • Compatible with HPLC systems and potentially UPLC platforms.
  • Manufactured in cGMP, ISO 9001:2000 certified plants.
Catalog No. 50-781-992