Learn More
Waters Corp Waters Atlantis T3 Column, C18, 100Å, 5 μm, 2.1 mm, 150 mm
Catalog No. 50781999 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Atlantis T3 columns are universal, silica-based, reversed-phase C18 columns for balanced retention of polar and non-polar compounds. Ideal for HPLC and UPLC separations, they feature advanced T3 bonding for enhanced polar chemical retention, aqueous compatibility, and extended low pH life.
  • Universal, silica-based, reversed-phase C18 column.
  • Balanced retention for polar and hydrophobic molecules.
  • Advanced T3 bonding technique with trifunctional C18 alkyl phase.
  • Patented T3 end-capping for improved efficiency.
  • Enhanced polar component retention and aqueous compatibility.
  • Improved column longevity, peak shape, and stability.
  • Extended life at low pH.
  • Better peak shape for bases containing amines at pH 7.
  • Performs over a wide pH range (2-8 pH).
  • Good retention and selectivity for neutral, hydrophobic, and polar analytes.
Catalog No. 50-781-999 Supplier Waters Corp Supplier No. 186003736
Atlantis T3 columns are universal, silica-based, reversed-phase C18 columns for balanced retention of polar and non-polar compounds. Ideal for HPLC and UPLC separations, they feature advanced T3 bonding for enhanced polar chemical retention, aqueous compatibility, and extended low pH life.
  • Universal, silica-based, reversed-phase C18 column.
  • Balanced retention for polar and hydrophobic molecules.
  • Advanced T3 bonding technique with trifunctional C18 alkyl phase.
  • Patented T3 end-capping for improved efficiency.
  • Enhanced polar component retention and aqueous compatibility.
  • Improved column longevity, peak shape, and stability.
  • Extended life at low pH.
  • Better peak shape for bases containing amines at pH 7.
  • Performs over a wide pH range (2-8 pH).
  • Good retention and selectivity for neutral, hydrophobic, and polar analytes.