Learn More
Waters Corp Waters Column, Atlantis T3, 5 μm, 3 mm X 50 mm
Catalog No. 50782001 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Atlantis T3 columns are universal, silica-based, reversed-phase C18 columns that provide balanced retention of polar and hydrophobic molecules. They are an excellent choice for developing separations of both polar and non-polar compounds.
  • Silica particle substrate for reversed-phase separation.
  • Optimized for balanced retention of polar and hydrophobic molecules.
  • pH range from 2 to 8.
  • Maximum pressure of 6000 psi (415 Bar).
  • T3 bonding technology.
  • Spherical particle shape.
  • 100 Å pore size.
  • 3 mm inner diameter and 50 mm length.
  • 14 % carbon load.
  • Designed for HPLC systems.
Catalog No. 50-782-001 Supplier Waters Corp Supplier No. 186003738
Add to Cart
Add to Cart
Atlantis T3 columns are universal, silica-based, reversed-phase C18 columns that provide balanced retention of polar and hydrophobic molecules. They are an excellent choice for developing separations of both polar and non-polar compounds.
  • Silica particle substrate for reversed-phase separation.
  • Optimized for balanced retention of polar and hydrophobic molecules.
  • pH range from 2 to 8.
  • Maximum pressure of 6000 psi (415 Bar).
  • T3 bonding technology.
  • Spherical particle shape.
  • 100 Å pore size.
  • 3 mm inner diameter and 50 mm length.
  • 14 % carbon load.
  • Designed for HPLC systems.