Learn More
Waters Corp Waters Atlantis T3 Kit, 100Å, 3 μm, 4.6 x 150 mm
Catalog No. 50782012 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Atlantis T3 columns are universal, silica-based, reversed-phase C18 columns providing balanced retention of polar and hydrophobic molecules. They are ideal for developing separations of polar and non-polar compounds. This Method Validation Kit includes three columns.
  • Utilizes T3 bonding with a trifunctional C18 alkyl phase for enhanced polar chemical retention and mobile phase compatibility.
  • Features efficient T3 end-capping, providing improved polar compound retention, aqueous compatibility, and column performance.
  • Delivers extended column life under low pH conditions by preventing silica substrate dissolution, improving longevity and peak shape.
  • Facilitates translation of HPLC techniques to UPLC methods, though some optimization may be needed.
Catalog No. 50-782-012 Supplier Waters Corp Supplier No. 186003751
Atlantis T3 columns are universal, silica-based, reversed-phase C18 columns providing balanced retention of polar and hydrophobic molecules. They are ideal for developing separations of polar and non-polar compounds. This Method Validation Kit includes three columns.
  • Utilizes T3 bonding with a trifunctional C18 alkyl phase for enhanced polar chemical retention and mobile phase compatibility.
  • Features efficient T3 end-capping, providing improved polar compound retention, aqueous compatibility, and column performance.
  • Delivers extended column life under low pH conditions by preventing silica substrate dissolution, improving longevity and peak shape.
  • Facilitates translation of HPLC techniques to UPLC methods, though some optimization may be needed.