Learn More
Waters Corp Atlantis T3 Kit, C18, 100 Å, 5 μm
Supplier: Waters Corp 186003754
The Waters Atlantis Silica T3 Method Validation Kit contains Atlantis T3 columns, which are universal, silica-based, reversed-phase C18 columns. These columns are engineered for balanced retention of polar and hydrophobic molecules and are fully compatible with 100% aqueous mobile phases.
- Silica-based, reversed-phase C18 columns.
- Designed for retention and separation of small, water-soluble polar organic compounds.
- Provides balanced retention for broad analyte mixtures.
- Compatible with 100% aqueous mobile phases.
- Eliminates the need for ion-pairing reagents, improving detection limits, method reproducibility, and robustness.
- Offers exceptional column lifetime at low pH.
- Columns have a 100 Å pore size and 5 μm particle size.
- T3 bonding ensures improved column lifetimes and reproducible performance.
- Compatible with both HPLC and UHPLC systems.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.