Learn More
Waters Corp Waters Corporation Sentry Guard Cartridge, C18, 3 μm, 2.1 mm, 10 mm
Catalog No. 50782015 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Atlantis T3 columns are universal, silica-based, reversed-phase C18 columns that provide balanced retention of polar and hydrophobic molecules. These columns are designed to function well within a pH range of 2 to 8 and can withstand a maximum pressure of 6,000 psi (415 Bar).
  • Universal, silica-based, reversed-phase C18 columns
  • Provides balanced retention of polar and hydrophobic molecules
  • Excellent choice for developing separations of polar and non-polar compounds
  • Functions well within the 2 to 8 pH range
  • Withstands a maximum of 6,000 psi (415 Bar) pressure
Catalog No. 50-782-015 Supplier Waters Corp Supplier No. 186003756
Atlantis T3 columns are universal, silica-based, reversed-phase C18 columns that provide balanced retention of polar and hydrophobic molecules. These columns are designed to function well within a pH range of 2 to 8 and can withstand a maximum pressure of 6,000 psi (415 Bar).
  • Universal, silica-based, reversed-phase C18 columns
  • Provides balanced retention of polar and hydrophobic molecules
  • Excellent choice for developing separations of polar and non-polar compounds
  • Functions well within the 2 to 8 pH range
  • Withstands a maximum of 6,000 psi (415 Bar) pressure