Learn More
Waters Corp Waters Atlantis Sentry Guard Cartridge, 100Å, 5 μm, 4.6 mm × 20 mm
Catalog No. 50782020 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Sentry Guard Cartridge protects analytical columns by removing chemical impurities and suspended solids. Utilizing universal, silica-based, reversed-phase C18 chemistry, it ensures balanced retention for polar and hydrophobic molecules. Designed for direct attachment, it minimizes extra column volumes and reduces leaks. Manufactured in a cGMP, ISO 9001 certified plant, each cartridge is tested for performance.
  • Protects compatible analytical columns
  • Universal, silica-based, reversed-phase C18 chemistry
  • Provides balanced retention for polar and hydrophobic molecules
  • Removes chemical impurities and suspended solids
  • Single unit design for direct attachment to the analytical column
  • Minimizes extra column volumes and reduces leaking
  • Manufactured in a cGMP, ISO 9001 certified plant
  • Each batch chromatographically tested and certified for use
Catalog No. 50-782-020 Supplier Waters Corp Supplier No. 186003761
This Sentry Guard Cartridge protects analytical columns by removing chemical impurities and suspended solids. Utilizing universal, silica-based, reversed-phase C18 chemistry, it ensures balanced retention for polar and hydrophobic molecules. Designed for direct attachment, it minimizes extra column volumes and reduces leaks. Manufactured in a cGMP, ISO 9001 certified plant, each cartridge is tested for performance.
  • Protects compatible analytical columns
  • Universal, silica-based, reversed-phase C18 chemistry
  • Provides balanced retention for polar and hydrophobic molecules
  • Removes chemical impurities and suspended solids
  • Single unit design for direct attachment to the analytical column
  • Minimizes extra column volumes and reduces leaking
  • Manufactured in a cGMP, ISO 9001 certified plant
  • Each batch chromatographically tested and certified for use