Learn More
Waters Corp Waters Prep Guard Cartridge, 5 μm, 100 Å, 10 mm X 10 mm
Catalog No. 50781958 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Atlantis T3 Prep Guard Cartridge is a universal, silica-based, reversed-phase C18 column designed for the balanced retention of polar and hydrophobic molecules. It is an ideal choice for developing separations of polar and non-polar compounds in HPLC and LC/MS applications.
  • Universal, silica-based, reversed-phase C18 chemistry
  • Provides balanced retention of polar and hydrophobic molecules
  • Ideal for developing separations of polar and non-polar compounds
  • Features a particle size of 5 μm and a pore size of 100 Å
  • Operates within a pH range of 2 to 8
  • Withstands temperatures up to 45 °C and pressures up to 6000 psi (415 Bar)
  • Endcapped with T3 bonding technology
Catalog No. 50-781-958 Supplier Waters Corp Supplier No. 186003695
This Atlantis T3 Prep Guard Cartridge is a universal, silica-based, reversed-phase C18 column designed for the balanced retention of polar and hydrophobic molecules. It is an ideal choice for developing separations of polar and non-polar compounds in HPLC and LC/MS applications.
  • Universal, silica-based, reversed-phase C18 chemistry
  • Provides balanced retention of polar and hydrophobic molecules
  • Ideal for developing separations of polar and non-polar compounds
  • Features a particle size of 5 μm and a pore size of 100 Å
  • Operates within a pH range of 2 to 8
  • Withstands temperatures up to 45 °C and pressures up to 6000 psi (415 Bar)
  • Endcapped with T3 bonding technology