Learn More
Waters Corp Waters Corporation Prep Guard Cartridge, 5 μm, 19 mm × 10 mm
Catalog No. 50781962 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This silica-based, reversed-phase C18 guard cartridge protects analytical columns and maintains optimal performance. It offers balanced retention for polar and hydrophobic molecules, making it ideal for separations of various compound types.
  • Constructed from the same packing material as the analytical column for compatibility.
  • Inserted between the injector and analytical column to prevent impurities and suspended solids.
  • Designed for direct attachment to the column, minimizing excess volumes and reducing leaks.
  • Manufactured in a cGMP, ISO 9001 certified plant using ultra-pure chemicals.
  • Each batch of packing material undergoes chromatographic evaluation with acidic, basic, and neutral analytes.
  • Validated and verified to ensure optimal performance, enhancing selectivity, sensitivity, and speed of studies.
  • Operational up to a maximum of 45 °C.
Catalog No. 50-781-962 Supplier Waters Corp Supplier No. 186003699
This silica-based, reversed-phase C18 guard cartridge protects analytical columns and maintains optimal performance. It offers balanced retention for polar and hydrophobic molecules, making it ideal for separations of various compound types.
  • Constructed from the same packing material as the analytical column for compatibility.
  • Inserted between the injector and analytical column to prevent impurities and suspended solids.
  • Designed for direct attachment to the column, minimizing excess volumes and reducing leaks.
  • Manufactured in a cGMP, ISO 9001 certified plant using ultra-pure chemicals.
  • Each batch of packing material undergoes chromatographic evaluation with acidic, basic, and neutral analytes.
  • Validated and verified to ensure optimal performance, enhancing selectivity, sensitivity, and speed of studies.
  • Operational up to a maximum of 45 °C.