Learn More
Waters Corp Waters Corporation Intelligent Speed Column, C18, 3 μm, 2.1 mm x 20 mm
Catalog No. 50781978 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Atlantis T3 columns are universal, silica-based, reversed-phase C18 columns that provide balanced retention of polar and hydrophobic molecules. They are recommended as a first choice when developing separations of polar and non-polar compounds.
  • C18 chemistry
  • Reversed phase separation mode
  • Silica particle substrate
  • pH range of 2 to 8
  • Maximum pressure of 6000 psi (415 Bar)
  • Endcapped
  • T3 bonding technology
  • Medium silanol activity
  • Spherical particle shape
  • 3 μm particle size
  • 100 Å pore size
  • Intelligent Speed (IS) column format
  • 330 surface area
  • HPLC system compatible
  • L1 USP classification
  • 2.1 mm inner diameter
  • 20 mm length
  • 14% carbon load
  • 1 unit per package
Catalog No. 50-781-978 Supplier Waters Corp Supplier No. 186003715
Atlantis T3 columns are universal, silica-based, reversed-phase C18 columns that provide balanced retention of polar and hydrophobic molecules. They are recommended as a first choice when developing separations of polar and non-polar compounds.
  • C18 chemistry
  • Reversed phase separation mode
  • Silica particle substrate
  • pH range of 2 to 8
  • Maximum pressure of 6000 psi (415 Bar)
  • Endcapped
  • T3 bonding technology
  • Medium silanol activity
  • Spherical particle shape
  • 3 μm particle size
  • 100 Å pore size
  • Intelligent Speed (IS) column format
  • 330 surface area
  • HPLC system compatible
  • L1 USP classification
  • 2.1 mm inner diameter
  • 20 mm length
  • 14% carbon load
  • 1 unit per package