Learn More
Waters Corp Atlantis Intelligent Speed Column, 100Å, 3 μm, 4.6 mm X 20 mm
Catalog No. 50781987 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Atlantis T3 columns are universal, silica-based, reversed-phase C18 columns designed for balanced retention of polar and hydrophobic molecules. They feature innovative T3 bonding and patented T3 end-capping for improved performance, aqueous compatibility, and a wider pH range.
  • Universal, silica-based, reversed-phase C18 column.
  • Balanced retention of polar and hydrophobic molecules.
  • Innovative T3 bonding for polar retention and water mobile phase compatibility.
  • Patented T3 end-capping for enhanced polar retention and aqueous compatibility.
  • Enhanced performance over a wider pH range.
  • Good retention and selectivity for diverse analytes.
  • Improved column lifetime, peak shape, and stability.
  • Better peak shapes for bases at pH 7.
  • Longer lifetimes at low pH.
  • Separates highly polar compounds effectively.
Catalog No. 50-781-987 Supplier Waters Corp Supplier No. 186003724
Add to Cart
Add to Cart
Atlantis T3 columns are universal, silica-based, reversed-phase C18 columns designed for balanced retention of polar and hydrophobic molecules. They feature innovative T3 bonding and patented T3 end-capping for improved performance, aqueous compatibility, and a wider pH range.
  • Universal, silica-based, reversed-phase C18 column.
  • Balanced retention of polar and hydrophobic molecules.
  • Innovative T3 bonding for polar retention and water mobile phase compatibility.
  • Patented T3 end-capping for enhanced polar retention and aqueous compatibility.
  • Enhanced performance over a wider pH range.
  • Good retention and selectivity for diverse analytes.
  • Improved column lifetime, peak shape, and stability.
  • Better peak shapes for bases at pH 7.
  • Longer lifetimes at low pH.
  • Separates highly polar compounds effectively.