Learn More
Waters Corp Atlantis T3 OBD Prep Column, 10 μm, 100 Å, 30 mm × 150 mm
Catalog No. 50781974 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These universal, silica-based, reversed-phase C18 columns ensure balanced retention for polar and hydrophobic molecules. Optimized for high retention and loading of highly polar substances, they facilitate efficient preparative runs and faster library screening. Featuring patented T3 end-capping, these columns offer superior analyte retention, excellent peak form, and low pH stability without ion-pairing reagents.
  • Balanced retention for polar and hydrophobic molecules.
  • High retention and loading for highly polar substances.
  • Facilitates efficient preparative runs and faster library screening.
  • Patented T3 end-capping for superior peak form and analyte retention.
  • Offers low pH stability without ion-pairing reagents.
Catalog No. 50-781-974 Supplier Waters Corp Supplier No. 186003711
Add to Cart
Add to Cart
Request a Quote
These universal, silica-based, reversed-phase C18 columns ensure balanced retention for polar and hydrophobic molecules. Optimized for high retention and loading of highly polar substances, they facilitate efficient preparative runs and faster library screening. Featuring patented T3 end-capping, these columns offer superior analyte retention, excellent peak form, and low pH stability without ion-pairing reagents.
  • Balanced retention for polar and hydrophobic molecules.
  • High retention and loading for highly polar substances.
  • Facilitates efficient preparative runs and faster library screening.
  • Patented T3 end-capping for superior peak form and analyte retention.
  • Offers low pH stability without ion-pairing reagents.