Learn More
Waters Corp Atlantis T3 OBD Prep Column, 100Å, 5 µm, 30 mm X 75 mm, 1/pk
Catalog No. 50781964 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Atlantis T3 columns are universal, silica-based, reversed-phase C18 columns that provide balanced retention of polar and hydrophobic molecules and should be your first choice when developing separations of polar and non-polar compounds.
Catalog No. 50-781-964 Supplier Waters Corp Supplier No. 186003701
Add to Cart
Add to Cart
Request a Quote
Atlantis T3 columns are universal, silica-based, reversed-phase C18 columns that provide balanced retention of polar and hydrophobic molecules and should be your first choice when developing separations of polar and non-polar compounds.