Learn More
Waters Corp Waters Atlantis T3 Column, 100Å, 5 μm, 10 mm x 150 mm
Catalog No. 502224197 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Atlantis T3 columns are universal, silica-based, reversed-phase C18 columns designed for balanced retention of both polar and hydrophobic molecules, making them a primary choice for developing separations of polar and non-polar compounds.
  • Provides balanced retention of both polar and hydrophobic molecules.
  • Primary choice for developing separations of polar and non-polar compounds.
  • Universal, silica-based, reversed-phase C18 columns.
Catalog No. 50-222-4197 Supplier Waters Corp Supplier No. 186008204
Atlantis T3 columns are universal, silica-based, reversed-phase C18 columns designed for balanced retention of both polar and hydrophobic molecules, making them a primary choice for developing separations of polar and non-polar compounds.
  • Provides balanced retention of both polar and hydrophobic molecules.
  • Primary choice for developing separations of polar and non-polar compounds.
  • Universal, silica-based, reversed-phase C18 columns.