Learn More
Waters Corp Waters Atlantis Silica T3 VanGuard Cartridge, 100Å, 3 μm, 2.1 mm x 5 mm
Catalog No. 502038150 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This VanGuard Cartridge extends analytical column lifetime and performance by filtering particulate and chemical contamination from the mobile phase stream. It safeguards 2.1 mm and 3.0 mm I.D. analytical columns using 3 μm sorbent particles.
  • Extends analytical column lifetime and performance.
  • Removes particulate contamination from the mobile phase stream.
  • Optimized for 2.1 mm and 3.0 mm I.D. analytical columns with 3 μm sorbent particles.
  • Compatible with analytical columns for reversed-phase Liquid Chromatography.
  • Manufactured under strict cGMP, ISO 9001 conditions.
  • Tested and certified for optimal performance.
  • Maintains UPLC, UHPLC, and HPLC separation efficiency.
  • Tolerates pressures up to 18000 psi (1240 bar).
  • Works across a wide pH range of 2 to 8.
Catalog No. 50-203-8150 Supplier Waters Corp Supplier No. 186007674
This VanGuard Cartridge extends analytical column lifetime and performance by filtering particulate and chemical contamination from the mobile phase stream. It safeguards 2.1 mm and 3.0 mm I.D. analytical columns using 3 μm sorbent particles.
  • Extends analytical column lifetime and performance.
  • Removes particulate contamination from the mobile phase stream.
  • Optimized for 2.1 mm and 3.0 mm I.D. analytical columns with 3 μm sorbent particles.
  • Compatible with analytical columns for reversed-phase Liquid Chromatography.
  • Manufactured under strict cGMP, ISO 9001 conditions.
  • Tested and certified for optimal performance.
  • Maintains UPLC, UHPLC, and HPLC separation efficiency.
  • Tolerates pressures up to 18000 psi (1240 bar).
  • Works across a wide pH range of 2 to 8.