Learn More
AGI NORTH AMERICA INC ATLAS SYRINGE PUMP VALVES
SDP
Catalog No. NC3456363
Encompass
  • Enhanced chemical compatibility with PTFE and PCTFE materials.
  • 4-port design for versatile fluidic control.
  • User-changeable ports for flexible configuration.
  • Robust integration with the Atlas XL Syringe Pump.
  • Optimizes volumetric dosing and sampling.
  • Suitable for aggressive chemical applications.
  • Individual port configuration as input or output.
Catalog No. NC3456363 Supplier AGI NORTH AMERICA INC Supplier No. 2200384
  • Enhanced chemical compatibility with PTFE and PCTFE materials.
  • 4-port design for versatile fluidic control.
  • User-changeable ports for flexible configuration.
  • Robust integration with the Atlas XL Syringe Pump.
  • Optimizes volumetric dosing and sampling.
  • Suitable for aggressive chemical applications.
  • Individual port configuration as input or output.