Learn More
Biotage Atlantic C-18 Disks, 47 mm
SDP
Catalog No. NC0196673
Encompass
Biotage is the leader in disk SPE technology. Atlantic SPE Disks are the only Certified for Automation disks available on the market today. Biotage “Certified for Automation”, Atlantic SPE Disks are QC-tested in our lab to guarantee that you receive the highest quality results when used in conjunction with the Biotage Horizon 5000 system. This silica based C18 is end-capped to eliminate unpredictable secondary interactions with unreacted silanols. This reversed-phase media can be used for EPA Method 525.2 along with other drinking and wastewater methods. See the SPE Disk Guide for a more detailed description matching disk-types and the compounds for which they may be used.
Catalog No. NC0196673 Supplier Biotage Supplier No. 47234602
Biotage is the leader in disk SPE technology. Atlantic SPE Disks are the only Certified for Automation disks available on the market today. Biotage “Certified for Automation”, Atlantic SPE Disks are QC-tested in our lab to guarantee that you receive the highest quality results when used in conjunction with the Biotage Horizon 5000 system. This silica based C18 is end-capped to eliminate unpredictable secondary interactions with unreacted silanols. This reversed-phase media can be used for EPA Method 525.2 along with other drinking and wastewater methods. See the SPE Disk Guide for a more detailed description matching disk-types and the compounds for which they may be used.