Learn More
Eurofins Discoverx ATM, active, 250 µg. (14-933M)
Supplier:  Eurofins Discoverx 14933M
Encompass
Recombinant, human FLAG-tagged ATM full length. ATM is a serine/threonine protein kinase which activates checkpoint signaling upon double strand breaks (DSBs), apoptosis and genotoxic stresses such as ionizing UV A light, thereby acting as a DNA damage sensor. Also plays a role in pre-B cell allelic exclusion, a process leading to expression of a single Ig heavy chain allele to enforce clonality and monospecific recognition by the B-cell antigen receptor (BCR) expressed on individual B-lymphocytes. Also involved in signal transduction and cell cycle control. Necessary for activation of ABL1 and SAPK. Phosphorylates DYRK2, CHEK2, p53/TP53, FANCD2, NFKBIA, BRCA1, CTIP, nibrin (NBN), TERF1, RAD9 and DCLRE1C. Phosphorylation of DYRK2 in nucleus in response to genotoxic stress prevents its MDM2-mediated ubiquitination and subsequent proteasome degradation. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9440
Add to Cart
Request a Quote