Learn More
Biomatik Corporation Atm Recombinant Antibody
Supplier:  Biomatik Corporation RAC07016
Encompass_Preferred
Atm Recombinant Antibody is a Human (Homo sapiens) Recombinant Antibody to ATM. The Atm Recombinant Antibody has been validated for the following applications: ELISA, WB, IHC, IP. Host: HEK293F Cell. Species: Human (Homo sapiens). Immunogen: A Synthesized Peptide Derived From Human Atm. Applications: ELISA, WB, IHC, IP. Target Synonyms: ATM. Accession Number: Q13315. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9269