Learn More
Cell Signaling Technology ATM Substrates Antibody Sampler Kit, Unconjugated, 1 Kit
SDP
Catalog No. 503133683
Encompass
The ATM Substrates Antibody Sampler Kit provides an economical means to evaluate the signaling of ATM to downstream substrates. This kit contains a selection of antibodies directed against key proteins involved in the DNA damage response, facilitating the analysis of ATM-mediated signaling pathways.
  • Includes primary antibodies for detection of p53, p95/NBS1, Chk2, and Rad50, along with their phosphorylated forms.
  • Contains an HRP-linked secondary antibody for convenient Western blot detection.
  • Offers sufficient volume to perform two Western blot experiments per primary antibody.
  • Provided in a stabilizing buffer with 50% glycerol for storage at –20°C.
Catalog No. 50-313-3683 Supplier Cell Signaling Technology Supplier No. 47833T
The ATM Substrates Antibody Sampler Kit provides an economical means to evaluate the signaling of ATM to downstream substrates. This kit contains a selection of antibodies directed against key proteins involved in the DNA damage response, facilitating the analysis of ATM-mediated signaling pathways.
  • Includes primary antibodies for detection of p53, p95/NBS1, Chk2, and Rad50, along with their phosphorylated forms.
  • Contains an HRP-linked secondary antibody for convenient Western blot detection.
  • Offers sufficient volume to perform two Western blot experiments per primary antibody.
  • Provided in a stabilizing buffer with 50% glycerol for storage at –20°C.