Learn More
Aviva Systems Biology Corp ATMIN Antibody - middle region (ARP32502_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32502_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ATMIN. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ATMIN. Alternate names: ASCIZ, DKFZp779K1455, FLJ76795, KIAA0431, znf822, ZNF822ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-54627