Learn More
Sigma Aldrich Fine Chemicals Biosciences Aldrich AtmosBag, Non-Sterile, Large, each
Catalog No. NC0776014
Encompass
AtmosBag glove bag is a flexible, inflatable polyethylene chamber with built-in gloves that lets you work in a totally isolated and controlled environment. It is an inexpensive, portable alternative to the glove box and is well suited to a variety of tasks and materials.
Catalog No. NC0776014 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z5302201EA
AtmosBag glove bag is a flexible, inflatable polyethylene chamber with built-in gloves that lets you work in a totally isolated and controlled environment. It is an inexpensive, portable alternative to the glove box and is well suited to a variety of tasks and materials.