Learn More
Andwin Scientific ATMOSBAG TWO-HAND MEDIUM
SDP
Catalog No. NC9092479
Encompass_Preferred
The Aldrich AtmosBag is a flexible and inflatable polyethylene chamber that can be used as a low-priced and mobile alternative to a glove box. It is able to cover top of cans and drums to isolate vapors and dusts throughout sampling procedures. This product can also act as a portable laboratory that can be used to administer tests, crime scene investigations, and other procedures wherein a clean isolated chamber is required. The bags also feature security with its seams that are heat-sealed.
Catalog No. NC9092479 Supplier Andwin Scientific Supplier No. Z118362
The Aldrich AtmosBag is a flexible and inflatable polyethylene chamber that can be used as a low-priced and mobile alternative to a glove box. It is able to cover top of cans and drums to isolate vapors and dusts throughout sampling procedures. This product can also act as a portable laboratory that can be used to administer tests, crime scene investigations, and other procedures wherein a clean isolated chamber is required. The bags also feature security with its seams that are heat-sealed.