Learn More
Waters Corp Atmospheric Pressure Photoionization Test Solution
Supplier:  Waters Corp 700002752
Encompass_Preferred
Mass Spectrometry standards kit for system assessment. 1 X 5mL Acetaminophen 1mg/L in 30% Methanol 70% Water 5mL amber flame sealed ampule Certificate of Analysis
Catalog No. 50-201-668