Learn More
Selleck Chemical LLC ATN-161
SDP
Supplier:  Selleck Chemical LLC S845425MG
Encompass_Preferred
ATN-161 is a novel small peptide antagonist of integrin 5 1 It binds to several integrins including 5 1 and v 3 that play a role in angiogenesis and tumor progression
Catalog No. 50-000-01713