Learn More
Selleck Chemical LLC ATN-161 Ac-PHSCN-NH2-5mg
SDP
Catalog No. 502389988
Encompass_Preferred
ATN-161 (Ac-PHSCN-NH2) is a novel small peptide antagonist of integrin α5β1. It binds to several integrins, including α5β1 and αvβ3, that play a role in angiogenesis and tumor progression.
Catalog No. 50-238-9988 Supplier Selleck Chemical LLC Supplier No. S84545MG
ATN-161 (Ac-PHSCN-NH2) is a novel small peptide antagonist of integrin α5β1. It binds to several integrins, including α5β1 and αvβ3, that play a role in angiogenesis and tumor progression.