Learn More
Medchemexpress LLC ATN-161 trifluoroacetate salt | 904763-27-5 | 99.0% | 711.67 g/mol | C25H36F3N9O10S | 5 MG
SDP
Catalog No. 5000223496
Encompass_Preferred
ATN-161 trifluoroacetate salt is a synthetic peptide integrin α5β1 antagonist supplied for research use. It inhibits angiogenesis and reduces liver metastasis growth in murine models. The trifluoroacetate salt is provided as a high-purity solid powder suitable for biochemical and in vivo studies.
  • Integrin α5β1 antagonist activity demonstrated in murine models.
  • High purity (99.0%) suitable for research applications.
  • Supplied as a white to off-white solid for easy handling and storage.
  • Available in small research pack sizes for dose-response and pilot studies.
  • Intended for laboratory research use only.
Catalog No. 50-002-23496 Supplier Medchemexpress LLC Supplier No. HY13535A5MG
ATN-161 trifluoroacetate salt is a synthetic peptide integrin α5β1 antagonist supplied for research use. It inhibits angiogenesis and reduces liver metastasis growth in murine models. The trifluoroacetate salt is provided as a high-purity solid powder suitable for biochemical and in vivo studies.
  • Integrin α5β1 antagonist activity demonstrated in murine models.
  • High purity (99.0%) suitable for research applications.
  • Supplied as a white to off-white solid for easy handling and storage.
  • Available in small research pack sizes for dose-response and pilot studies.
  • Intended for laboratory research use only.