Learn More
TARGETMOL CHEMICALS INC ATN-224 5MG
SDP
Catalog No. 502581295
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. ATN-224 (Bis(choline)tetrathiomolybdate) inhibits SOD1 activity in endothelial cells an effect that is dose dependent with an IC50 of 17.5±3.7 nM. ATN-224 is an oral Cu2+/Zn2+-superoxide dismutase 1 (SOD1) inhibitor. purity: 99%
Catalog No. 50-258-1295 Supplier TARGETMOL CHEMICALS INC Supplier No. T143415MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. ATN-224 (Bis(choline)tetrathiomolybdate) inhibits SOD1 activity in endothelial cells an effect that is dose dependent with an IC50 of 17.5±3.7 nM. ATN-224 is an oral Cu2+/Zn2+-superoxide dismutase 1 (SOD1) inhibitor. purity: 99%