Learn More
Cayman Chemical ATN-161 (trifluoroacetate salt), Purity: ≥95%, CAS: 904763-27-5, 10 mg
SDP
Catalog No. 502947665
Encompass_Preferred
An integrin receptor antagonist; binds to αVβ3 and αVβ5 receptors (Kds = 0.69 and 1 µM, respectively) in MDA-MB-231 cells; inhibits the interaction between αVβ5 and human ACE2 or SARS-CoV-2 spike protein in cell-free assays; reduces viral titers in Vero E6 cells infected with SARS-CoV-2 (EC50 = 3.16 µM); inhibits VEGF-induced migration and capillary tube formation of primary human choroidal endothelial cells at 0.1 µM; reduces tumor growth and metastasis in an MDA-MB-231 mouse xenograft model at 1 mg/kg; reduces infarct volume and edema in a mouse model of MCAO-induced ischemic stroke at 1 mg/kg
Catalog No. 50-294-7665 Supplier Cayman Chemical Supplier No. 3405610 MG
An integrin receptor antagonist; binds to αVβ3 and αVβ5 receptors (Kds = 0.69 and 1 µM, respectively) in MDA-MB-231 cells; inhibits the interaction between αVβ5 and human ACE2 or SARS-CoV-2 spike protein in cell-free assays; reduces viral titers in Vero E6 cells infected with SARS-CoV-2 (EC50 = 3.16 µM); inhibits VEGF-induced migration and capillary tube formation of primary human choroidal endothelial cells at 0.1 µM; reduces tumor growth and metastasis in an MDA-MB-231 mouse xenograft model at 1 mg/kg; reduces infarct volume and edema in a mouse model of MCAO-induced ischemic stroke at 1 mg/kg