Learn More
Aviva Systems Biology Corp ATOH1 Antibody - middle region (ARP32365_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32365_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ATOH1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ATOH1. Alternate names: ATH1, HATH1, MATH-1, bHLHa14ApplicationIHC, WB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-54546